Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080907.2 | NP_543183.1 | 1 | 167 | ||
GenBank mRNA | AF332004.1 | - | 1 | 85 | ||
BC061726.1 | - | 1 | 167 | 5598703 |