Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012137.3 | NP_036269.1 | 1, 2 | 465 | ||
GenBank mRNA | AK312432.1 | - | 1, 2 | 426 | ||
BC033680.1 | - | 1, 2 | 403 | 5189970 | ||
BC043235.2 | - | 1, 2 | 187 | |||
DC324361.1 | - | 1, 2 | 467 | |||
EU794605.1 | - | 1, 2 | 403 |