Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130432.2 | NP_569116.1 | 5, 6 | 448 | ||
GenBank mRNA | BC084724.1 | - | 5, 6 | 424 | 7318693 | |
FQ209976.1 | - | 5, 6 | 446 | |||
FQ209999.1 | - | 5, 6 | 445 | |||
FQ210203.1 | - | 5, 6 | 442 | |||
FQ210309.1 | - | 5, 6 | 441 | |||
FQ217627.1 | - | 5, 6 | 442 | |||
FQ220591.1 | - | 5, 6 | 440 | |||
FQ220785.1 | - | 5, 6 | 442 | |||
FQ221346.1 | - | 5, 6 | 442 | |||
FQ221555.1 | - | 5, 6 | 442 | |||
FQ221602.1 | - | 5, 6 | 444 | |||
FQ221859.1 | - | 5, 6 | 445 | |||
FQ222187.1 | - | 5, 6 | 442 | |||
FQ222266.1 | - | 5, 6 | 445 | |||
FQ222359.1 | - | 5, 6 | 442 | |||
FQ222425.1 | - | 4, 5 | 393 | |||
FQ222441.1 | - | 5, 6 | 445 | |||
FQ222573.1 | - | 5, 6 | 442 | |||
FQ222592.1 | - | 5, 6 | 442 | |||
FQ222643.1 | - | 5, 6 | 442 | |||
FQ222649.1 | - | 5, 6 | 442 | |||
FQ222885.1 | - | 5, 6 | 442 | |||
FQ223174.1 | - | 5, 6 | 442 | |||
FQ223351.1 | - | 5, 6 | 442 | |||
FQ223601.1 | - | 5, 6 | 442 | |||
FQ223978.1 | - | 5, 6 | 442 | |||
FQ224030.1 | - | 5, 6 | 442 | |||
FQ224170.1 | - | 5, 6 | 461 | |||
FQ225803.1 | - | 5, 6 | 440 | |||
FQ228324.1 | - | 5, 6 | 439 | |||
L01123.1 | - | 1, 2 | 51 | |||
X53378.1 | - | 5, 6 | 443 |
Gene Symbol: LOC683961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749925.3 | XP_003749973.2 | 1 | 453 |
Gene Symbol: LOC684988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001053043.4 | XP_001053043.1 | 1 | 434 |