Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130741.1 | NP_570097.1 | 2 | 271 | ||
GenBank mRNA | BC089053.1 | - | 2 | 333 | 7310185 | |
X13295.1 | - | 2 | 271 |