Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130894.4 | NP_570964.4 | 16 | 2177 | ||
XM_006239402.2 | XP_006239464.1 | 17 | 2330 | |||
XM_006239403.1 | XP_006239465.1 | 17 | 2359 | |||
XM_008764288.1 | XP_008762510.1 | 18 | 2347 | |||
GenBank mRNA | AB084165.1 | - | 15 | 2030 | ||
BC070880.1 | - | 16 | 2181 | 7096620 | ||
U41803.2 | - | 17 | 2195 |
Gene Symbol: LOC100911485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750086.3 | XP_003750134.1 | 2650 |