Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131907.2 | NP_571982.2 | 10, 11 | 1222 | ||
GenBank mRNA | BC086994.1 | - | 10, 11 | 1222 | 7106233 | |
M93017.1 | - | 10, 11 | 933 | |||
M93018.1 | - | 349 |