Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sep15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133297.2 | NP_579831.2 | 2, 3 | 527 | ||
GenBank mRNA | AF390544.1 | - | 2, 3 | 251 | ||
BC060547.1 | - | 2, 3 | 276 | 6921948 | ||
CK596377.1 | - | 2, 3 | 542 | |||
DY561321.1 | - | 2, 3 | 265 | |||
FQ209574.1 | - | 2, 3 | 262 | |||
FQ212727.1 | - | 2, 3 | 295 | |||
FQ213177.1 | - | 2, 3 | 276 | |||
FQ213838.1 | - | 2, 3 | 262 | |||
FQ214634.1 | - | 2, 3 | 272 | |||
FQ215362.1 | - | 2, 3 | 262 | |||
FQ215992.1 | - | 2, 3 | 285 | |||
FQ216831.1 | - | 2, 3 | 262 | |||
FQ219237.1 | - | 2, 3 | 283 | |||
FQ219570.1 | - | 2, 3 | 262 | |||
FQ219725.1 | - | 2, 3 | 255 | |||
FQ220311.1 | - | 2, 3 | 261 | |||
FQ220593.1 | - | 2, 3 | 267 | |||
FQ220877.1 | - | 2, 3 | 275 | |||
FQ229133.1 | - | 2, 3 | 268 | |||
FQ230131.1 | - | 2, 3 | 262 |