Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133519.4 | NP_598203.4 | 4, 5 | 545 | ||
GenBank mRNA | AF347935.1 | - | 4, 5 | 562 |