Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012162.3 | NP_036294.2 | 2 | 612 | ||
NM_024555.5 | NP_078831.4 | 2 | 612 | |||
GenBank mRNA | AF174592.1 | - | 2 | 269 | ||
AF370422.1 | - | 2 | 1608 | |||
AK026541.1 | - | 2 | 561 | |||
AK223088.1 | - | 2 | 561 | |||
AL713783.1 | - | 1 | 307 | |||
BC033066.1 | - | 2 | 601 | 5516945 | ||
BM761056.1 | - | 2 | 612 |