Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133581.1 | NP_598265.1 | 4 | 563 | ||
XM_006255705.2 | XP_006255767.1 | 4 | 816 | |||
GenBank mRNA | AF037272.1 | - | 4 | 563 | ||
BC063152.1 | - | 4 | 566 | 6887007 | ||
FQ213083.1 | - | 4 | 548 | |||
FQ216345.1 | - | 4 | 557 | |||
FQ216415.1 | - | 4 | 558 | |||
FQ219846.1 | - | 4 | 558 | |||
FQ219958.1 | - | 4 | 796 | |||
FQ220334.1 | - | 4 | 795 | |||
FQ229404.1 | - | 4 | 548 |