Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134349.3 | NP_599176.1 | 4 | 386 | ||
GenBank mRNA | BC063150.1 | - | 4 | 384 | 6922267 | |
FJ179404.1 | - | 3 | 315 | |||
FQ209427.1 | - | 4 | 407 | |||
FQ209464.1 | - | 4 | 399 | |||
FQ209465.1 | - | 4 | 406 | |||
FQ211049.1 | - | 4 | 406 | |||
FQ212841.1 | - | 4 | 407 | |||
FQ218096.1 | - | 4 | 389 | |||
FQ219010.1 | - | 4 | 408 | |||
FQ219464.1 | - | 4 | 412 | |||
FQ219581.1 | - | 4 | 408 | |||
FQ219850.1 | - | 4 | 398 | |||
J03752.1 | - | 4 | 370 |