Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134349.3 | NP_599176.1 | 4 | 699 | ||
GenBank mRNA | BC063150.1 | - | 4 | 697 | 6922267 | |
FQ209464.1 | - | 4 | 712 | |||
FQ211049.1 | - | 4 | 719 | |||
FQ212841.1 | - | 4 | 720 | |||
FQ218830.1 | - | 4 | 727 | |||
FQ219010.1 | - | 4 | 721 | |||
FQ219410.1 | - | 4 | 693 | |||
FQ219464.1 | - | 4 | 725 | |||
FQ219581.1 | - | 4 | 721 | |||
FQ219850.1 | - | 4 | 711 | |||
J03752.1 | - | 4 | 683 |