Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138548.2 | NP_612557.1 | 5 | 634 | ||
GenBank mRNA | D13374.1 | - | 5 | 635 |