Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138831.1 | NP_620186.1 | 2 | 644 | ||
XM_008772974.1 | XP_008771196.1 | 2 | 683 | |||
XM_008772975.1 | XP_008771197.1 | 2 | 682 | |||
GenBank mRNA | AB047324.1 | - | 2 | 644 |