Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138833.1 | NP_620188.1 | 2 | 310 | ||
XM_006244057.2 | XP_006244119.1 | 3 | 356 | |||
XM_006244058.2 | XP_006244120.1 | 3 | 390 | |||
XM_006244059.1 | XP_006244121.1 | 3 | 349 | |||
XM_006244060.2 | XP_006244122.1 | 2 | 364 | |||
XM_006244061.1 | XP_006244123.1 | 2 | 370 | |||
XM_006244062.2 | XP_006244124.1 | 2 | 298 | |||
XM_008766666.1 | XP_008764888.1 | 2 | 289 | |||
XM_008766667.1 | XP_008764889.1 | 2 | 431 | |||
GenBank mRNA | X89383.1 | - | 2 | 310 |