Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138909.1 | NP_620264.1 | 1 | 927 | ||
GenBank mRNA | Y11321.1 | - | 1 | 927 |