Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139060.3 | NP_620691.2 | 4, 5 | 877 | ||
XM_008768511.1 | XP_008766733.1 | 4, 5 | 882 | |||
GenBank mRNA | AB063114.1 | - | 4, 5 | 869 |