Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270788.1 | NP_001257717.1 | 4, 5 | 557 | ||
NM_139100.2 | NP_620800.2 | 4, 5 | 554 | |||
GenBank mRNA | BC070918.1 | - | 4, 5 | 496 | 7104964 | |
CB577607.1 | - | 4, 5 | 554 | |||
FQ210190.1 | - | 4, 5 | 508 | |||
FQ213750.1 | - | 4, 5 | 513 | |||
FQ214698.1 | - | 4, 5 | 511 | |||
FQ215315.1 | - | 4, 5 | 511 | |||
FQ215697.1 | - | 4, 5 | 511 | |||
FQ215957.1 | - | 4, 5 | 511 | |||
FQ216349.1 | - | 4, 5 | 510 | |||
FQ216361.1 | - | 4, 5 | 511 | |||
FQ216409.1 | - | 4, 5 | 518 | |||
FQ216421.1 | - | 4, 5 | 511 | |||
FQ217707.1 | - | 4, 5 | 511 | |||
FQ220537.1 | - | 4, 5 | 508 | |||
FQ220626.1 | - | 4, 5 | 495 | |||
FQ223916.1 | - | 4, 5 | 511 | |||
FQ227446.1 | - | 4, 5 | 508 | |||
FQ229113.1 | - | 4, 5 | 508 | |||
FQ230627.1 | - | 4, 5 | 508 | |||
FQ232293.1 | - | 2, 3 | 313 | |||
FQ232835.1 | - | 4, 5 | 504 | |||
FQ234854.1 | - | 4, 5 | 508 | |||
M23984.1 | - | 3, 4 | 473 |