Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prlhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139193.3 | NP_631932.3 | 1 | 621 | ||
GenBank mRNA | S77867.1 | - | 1 | 621 |
Set Membership: |
G-protein coupled receptor |