Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270364.1 | NP_001257293.1 | 3 | 400 | ||
NM_001270365.1 | NP_001257294.1 | 3 | 502 | |||
NM_012190.3 | NP_036322.2 | 3 | 502 | |||
NR_072979.1 | 3 | 502 | ||||
XM_006713481.2 | XP_006713544.1 | 3 | 241 | |||
XM_011512355.1 | XP_011510657.1 | 3 | 234 | |||
XM_017005613.1 | XP_016861102.1 | 3 | 625 | |||
XM_017005614.1 | XP_016861103.1 | 3 | 241 | |||
GenBank mRNA | AB209299.1 | - | 3 | 309 | ||
AF052732.1 | - | 3 | 262 | |||
AK222535.1 | - | 3 | 284 | |||
AK222694.1 | - | 3 | 284 | |||
AK294392.1 | - | 3 | 286 | |||
AK307699.1 | - | 4 | 413 | |||
AK308112.1 | - | 3 | 241 | |||
BC027241.1 | - | 3 | 275 | 4484301 | ||
BI829048.1 | - | 3 | 342 | |||
CR749807.1 | - | 3 | 400 | |||
DC342268.1 | - | 3 | 500 | |||
KM111188.1 | - | 3 | 311 | |||
KU178540.1 | - | 2 | 152 | |||
KU178541.1 | - | 3 | 336 | |||
KU178542.1 | - | 2 | 152 |