Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145770.2 | NP_665713.2 | 3 | 386 | ||
XM_006251743.2 | XP_006251805.1 | 4 | 471 | |||
XM_006251744.2 | XP_006251806.1 | 3 | 342 | |||
XM_006251745.2 | XP_006251807.1 | 4 | 389 | |||
GenBank mRNA | X95189.1 | - | 3 | 385 |