Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153301.2 | NP_695213.1 | 3, 4 | 488 | ||
GenBank mRNA | AF415240.1 | - | 3, 4 | 496 |