Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145667.1 | NP_001139139.1 | 1, 2 | 492 | ||
NM_012201.5 | NP_036333.2 | 1, 2 | 492 | |||
NR_027264.1 | 1, 2 | 492 | ||||
NR_027265.1 | 1, 2 | 492 | ||||
GenBank mRNA | AK027032.1 | - | 1, 2 | 453 | ||
BC031836.1 | - | 1, 2 | 446 | |||
BC060822.1 | - | 1, 2 | 463 | 5296427 | ||
DA493962.1 | - | 1, 2 | 492 | |||
U28811.1 | - | 1, 2 | 465 | |||
U64791.1 | - | 1, 2 | 445 |