Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172030.1 | NP_742027.1 | 2 | 227 | ||
XM_006233644.2 | XP_006233706.1 | 2 | 327 | |||
XM_008761606.1 | XP_008759828.1 | 2 | 327 | |||
GenBank mRNA | AF276940.1 | - | 2 | 227 | ||
BC086558.1 | - | 2 | 237 | 7302083 | ||
Y11835.1 | - | 2 | 226 |