Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012203.1 | NP_036335.1 | 4, 5 | 442 | ||
XM_011518073.2 | XP_011516375.1 | 4, 5 | 456 | |||
XM_017015320.1 | XP_016870809.1 | 4, 5 | 486 | |||
XM_017015321.1 | XP_016870810.1 | 4, 5 | 486 | |||
XM_017015323.1 | XP_016870812.1 | 4, 5 | 456 | |||
GenBank mRNA | AF113215.1 | - | 4, 5 | 451 | ||
AF134895.1 | - | 4, 5 | 442 | |||
AF146018.1 | - | 4, 5 | 441 | |||
AK307325.1 | - | 4, 5 | 428 | |||
AK307902.1 | - | 4, 5 | 400 | |||
AK315690.1 | - | 4, 5 | 456 | |||
BC000605.2 | - | 4, 5 | 421 | 3347232 | ||
BC003131.1 | - | 4, 5 | 442 | |||
BC088360.1 | - | 4, 5 | 440 |