Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173141.1 | NP_775164.1 | 2, 3 | 327 | ||
GenBank mRNA | AJ428954.1 | - | 2, 3 | 327 |