Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004439.1 | NP_001004439.1 | 5 | 504 | ||
XM_005254228.3 | XP_005254285.1 | 3 | 726 | |||
GenBank mRNA | AF109681.1 | - | 5 | 489 | ||
AF137378.2 | - | 5 | 507 | |||
AK095863.1 | - | 6 | 516 | |||
AK292445.1 | - | 5 | 513 | |||
AK304778.1 | - | 5 | 514 | |||
Z50167.1 | - | 3 | 210 |