Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178106.2 | NP_835207.1 | 4 | 424 | ||
XM_006244097.2 | XP_006244159.1 | 5 | 784 | |||
GenBank mRNA | AJ437217.1 | - | 4 | 424 |