Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dfnb31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181088.1 | NP_851602.1 | 6 | 1212 | ||
GenBank mRNA | AY227205.1 | - | 6 | 1212 |