Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012234.6 | NP_036366.3 | 1 | 258 | ||
GenBank mRNA | AB029551.1 | - | 1 | 267 | ||
AF179286.1 | - | 1 | 142 | |||
AF227959.1 | - | 1 | 73 | |||
AY228125.1 | - | 1 | 132 | |||
BC014959.1 | - | 1 | 226 | 4902201 | ||
BC036459.1 | - | 1 | 263 | 4813939 |