Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270871.1 | NP_001257800.1 | 15 | 1296 | ||
NM_001270872.1 | NP_001257801.1 | 14 | 1260 | |||
NM_181366.2 | NP_852031.1 | 16 | 1344 | |||
XM_006256916.2 | XP_006256978.1 | 16 | 1384 | |||
XM_008773154.1 | XP_008771376.1 | 15 | 1724 | |||
XM_008773155.1 | XP_008771377.1 | 15 | 1334 | |||
XM_008773156.1 | XP_008771378.1 | 14 | 1688 | |||
XM_008773157.1 | XP_008771379.1 | 14 | 1682 | |||
XM_008773158.1 | XP_008771380.1 | 13 | 1646 | |||
XM_008773159.1 | XP_008771381.1 | 12 | 1237 | |||
XM_008773160.1 | XP_008771382.1 | 15 | 1725 | |||
GenBank mRNA | AF538953.1 | - | 16 | 1354 | ||
AF538958.1 | - | 15 | 1306 | |||
AF538959.1 | - | 14 | 1270 |
Set Membership: |
G-protein coupled receptor |