Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC299282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182474.1 | NP_872280.1 | 3 | 675 | ||
GenBank mRNA | AY298742.1 | - | 3 | 675 | ||
AY325133.1 | - | 3 | 675 | |||
BC088096.1 | - | 3 | 999 | 7372979 | ||
D00752.1 | - | 3 | 982 | |||
FQ209483.1 | - | 3 | 995 | |||
FQ210703.1 | - | 3 | 997 | |||
FQ211200.1 | - | 1 | 70 | |||
FQ218472.1 | - | 3 | 1001 | |||
M15917.1 | - | 2 | 699 | |||
X16357.1 | - | 3 | 995 |