Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC299282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182474.1 | NP_872280.1 | 4 | 825 | ||
GenBank mRNA | AY298742.1 | - | 4 | 825 | ||
AY325133.1 | - | 4 | 825 | |||
BC088096.1 | - | 4 | 1149 | 7372979 | ||
D00752.1 | - | 4 | 1132 | |||
FQ209483.1 | - | 4 | 1145 | |||
FQ210703.1 | - | 4 | 1147 | |||
FQ211200.1 | - | 2 | 220 | |||
FQ218472.1 | - | 4 | 1151 | |||
FQ219154.1 | - | 2 | 911 | |||
FQ219226.1 | - | 5 | 1030 | |||
FQ219622.1 | - | 4 | 874 | |||
M15917.1 | - | 3 | 849 | |||
X16357.1 | - | 4 | 1145 |