Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myocd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182667.2 | NP_872608.1 | 13 | 2988 | ||
XM_006246582.2 | XP_006246644.1 | 14 | 5573 | |||
XM_006246583.2 | XP_006246645.1 | 15 | 5617 |