Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012249.3 | NP_036381.2 | 2, 3 | 528 | ||
XM_011532726.2 | XP_011531028.1 | 2, 3 | 705 | |||
XM_011532728.2 | XP_011531030.1 | 1, 2 | 94 | |||
XM_017003716.1 | XP_016859205.1 | 2, 3 | 847 | |||
GenBank mRNA | AF498976.1 | - | 2, 3 | 233 | ||
AK095329.1 | - | 2, 3 | 255 | |||
BC056154.1 | - | 2, 3 | 321 | 5539023 | ||
BC065291.1 | - | 2, 3 | 298 | 6061807 | ||
BC070485.1 | - | 2, 3 | 306 | 6010516 | ||
BC093805.1 | - | 2, 3 | 242 | 7939650 | ||
BC101806.1 | - | 2, 3 | 242 | 8069312 | ||
BU928962.1 | - | 1 | 523 | |||
EU794657.1 | - | 2, 3 | 528 | |||
M31470.1 | - | 2, 3 | 233 |