Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194461.1 | NP_919443.1 | 1 | 582 | ||
XM_006247055.2 | XP_006247117.1 | 2 | 747 | |||
XM_006247056.2 | XP_006247118.1 | 2 | 670 | |||
XM_006247057.2 | XP_006247119.1 | 2 | 652 |