Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111294.1 | NP_001104764.1 | 4 | 549 | ||
NM_001111295.1 | NP_001104765.1 | 4 | 483 | |||
NM_198132.3 | NP_937765.1 | 4 | 549 | |||
XM_008761911.1 | XP_008760133.1 | 4 | 498 | |||
XM_008761912.1 | XP_008760134.1 | 4 | 498 | |||
XM_008761913.1 | XP_008760135.1 | 4 | 544 | |||
GenBank mRNA | AY363226.1 | - | 4 | 495 | ||
AY363227.1 | - | 4 | 429 | |||
BC081878.1 | - | 4 | 489 | 7108128 | ||
CB585714.1 | - | 4 | 561 | |||
CB725005.1 | - | 4 | 421 | |||
FQ214617.1 | - | 4 | 445 |