Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198742.2 | NP_942037.2 | 2, 3 | 354 | ||
XM_006232507.1 | XP_006232569.1 | 2, 3 | 354 | |||
XM_008761084.1 | XP_008759306.1 | 3, 4 | 580 | |||
GenBank mRNA | AY365022.1 | - | 2, 3 | 180 | ||
BC081890.1 | - | 2, 3 | 354 | 7109511 | ||
FQ211230.1 | - | 2, 3 | 412 | |||
FQ218772.1 | - | 3, 4 | 585 |