Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198750.2 | NP_942045.2 | 3 | 853 | ||
XM_008765225.1 | XP_008763447.1 | 3 | 877 | |||
GenBank mRNA | AF545855.1 | - | 3 | 417 | ||
BC107677.1 | - | 3 | 853 | 7939194 |