Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198752.3 | NP_942047.2 | 7 | 776 | ||
GenBank mRNA | AJ250329.1 | - | 7 | 776 | ||
AJ250330.1 | - | 7 | 706 | |||
FQ211833.1 | - | 6 | 630 |