Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198753.2 | NP_942048.1 | 2, 3 | 224 | ||
GenBank mRNA | BC058494.1 | - | 2, 3 | 227 | 6887923 | |
FQ209477.1 | - | 2, 3 | 224 | |||
FQ211542.1 | - | 2, 3 | 224 | |||
FQ211764.1 | - | 2, 3 | 224 | |||
FQ212671.1 | - | 2, 3 | 224 | |||
FQ213017.1 | - | 2, 3 | 224 | |||
FQ213135.1 | - | 2, 3 | 226 | |||
FQ214177.1 | - | 2, 3 | 224 | |||
FQ214350.1 | - | 2, 3 | 224 | |||
FQ214687.1 | - | 2, 3 | 221 | |||
FQ215305.1 | - | 2, 3 | 224 | |||
FQ218147.1 | - | 2, 3 | 224 | |||
FQ219083.1 | - | 2, 3 | 224 | |||
FQ219302.1 | - | 2, 3 | 224 | |||
FQ219523.1 | - | 2, 3 | 223 | |||
FQ220029.1 | - | 2, 3 | 223 | |||
FQ220099.1 | - | 2, 3 | 224 | |||
FQ220378.1 | - | 2, 3 | 224 | |||
FQ220408.1 | - | 2, 3 | 220 | |||
FQ220423.1 | - | 2, 3 | 223 | |||
FQ220830.1 | - | 2, 3 | 224 | |||
FQ222720.1 | - | 2, 3 | 224 | |||
FQ222993.1 | - | 2, 3 | 224 | |||
FQ223514.1 | - | 2, 3 | 224 | |||
FQ225290.1 | - | 2, 3 | 224 | |||
FQ225405.1 | - | 2, 3 | 224 | |||
FQ225635.1 | - | 2, 3 | 224 | |||
FQ226331.1 | - | 2, 3 | 224 | |||
FQ227897.1 | - | 2, 3 | 224 | |||
FQ228916.1 | - | 2, 3 | 224 | |||
FQ229153.1 | - | 2, 3 | 224 | |||
FQ229505.1 | - | 2, 3 | 224 | |||
FQ229795.1 | - | 2, 3 | 220 | |||
FQ229869.1 | - | 2, 3 | 224 | |||
FQ229906.1 | - | 2, 3 | 224 | |||
FQ231551.1 | - | 2, 3 | 220 | |||
FQ232038.1 | - | 2, 3 | 224 | |||
FQ232101.1 | - | 2, 3 | 224 | |||
FQ233196.1 | - | 2, 3 | 224 | |||
FQ234897.1 | - | 2, 3 | 224 | |||
FQ235165.1 | - | 2, 3 | 224 | |||
X62166.1 | - | 2, 3 | 228 |