Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033674.1 | NP_001028846.1 | 1 | 205 | ||
NM_198765.2 | NP_942060.1 | 1 | 185 | |||
GenBank mRNA | AJ250312.1 | - | 1 | 185 | ||
BC100660.1 | - | 1 | 205 | 7935996 |