Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033674.1 | NP_001028846.1 | 7 | 4628 | ||
NM_198765.2 | NP_942060.1 | 8 | 2873 | |||
GenBank mRNA | AJ250312.1 | - | 8 | 2873 | ||
AJ250313.1 | - | 4 | 3947 | |||
BC100660.1 | - | 7 | 4628 | 7935996 |