Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198783.1 | NP_942078.1 | 1, 2 | 171 | ||
GenBank mRNA | AF283900.1 | - | 1, 2 | 171 |