Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198786.2 | NP_942081.2 | 3 | 592 | ||
GenBank mRNA | AY079080.1 | - | 3 | 452 | ||
AY079081.2 | - | 2 | 705 | |||
BC062388.1 | - | 3 | 592 | 5620891 |
Gene Symbol: LOC100911027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748700.3 | XP_003748748.1 | 3 | 549 |