Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199082.2 | NP_954513.2 | 2 | 191 | ||
XM_006247879.2 | XP_006247941.1 | 2 | 287 | |||
XM_006247880.2 | XP_006247942.1 | 3 | 1111 | |||
XM_006247881.2 | XP_006247943.1 | 2 | 828 | |||
XM_006247882.2 | XP_006247944.1 | 2 | 284 | |||
XM_006247884.2 | XP_006247946.1 | 3 | 1109 | |||
GenBank mRNA | BC061794.1 | - | 2 | 186 | 5597864 | |
CB544874.1 | - | 2 | 206 | |||
DY309177.1 | - | 1 | 279 |