Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199082.2 | NP_954513.2 | 3 | 371 | ||
XM_006247879.2 | XP_006247941.1 | 3 | 467 | |||
XM_006247880.2 | XP_006247942.1 | 4 | 1291 | |||
XM_006247881.2 | XP_006247943.1 | 3 | 1008 | |||
XM_006247882.2 | XP_006247944.1 | 3 | 464 | |||
XM_006247884.2 | XP_006247946.1 | 4 | 1289 | |||
GenBank mRNA | BC061794.1 | - | 3 | 366 | 5597864 | |
DY309177.1 | - | 2 | 459 |