Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199090.2 | NP_954521.1 | 5 | 823 | ||
XM_006230760.2 | XP_006230822.1 | 4 | 725 | |||
GenBank mRNA | BC060593.1 | - | 5 | 824 | 6887766 | |
FQ212766.1 | - | 5 | 770 | |||
FQ215165.1 | - | 5 | 751 | |||
FQ219356.1 | - | 5 | 774 |