Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199092.1 | NP_954523.1 | 3, 4 | 255 | ||
XM_006234150.2 | XP_006234212.1 | 5, 6 | 419 | |||
XM_006234151.2 | XP_006234213.1 | 4, 5 | 345 | |||
XM_006234152.2 | XP_006234214.1 | 6, 7 | 515 | |||
XM_006234154.2 | XP_006234216.1 | 3, 4 | 597 | |||
XM_008761704.1 | XP_008759926.1 | 3, 4 | 597 | |||
XM_008761705.1 | XP_008759927.1 | 5, 6 | 397 | |||
XM_008761706.1 | XP_008759928.1 | 4, 5 | 323 | |||
XM_008761707.1 | XP_008759929.1 | 4, 5 | 715 | |||
XM_008761708.1 | XP_008759930.1 | 4, 5 | 323 | |||
GenBank mRNA | BC060516.1 | - | 3, 4 | 255 | 5598062 | |
FQ213311.1 | - | 379 |