Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321441.1 | NP_001308370.1 | 2 | 205 | ||
NM_012111.2 | NP_036243.1 | 2 | 339 | |||
GenBank mRNA | AF164791.1 | - | 2 | 245 | ||
AJ243310.1 | - | 2 | 310 | |||
AK300766.1 | - | 2 | 337 | |||
AK313824.1 | - | 2 | 273 | |||
BC000321.2 | - | 2 | 287 | 2821759 | ||
BG432594.1 | - | 2 | 340 | |||
BQ073652.1 | - | 2 | 129 | |||
CX786861.1 | - | 2 | 216 |